siRNA Details
Gene Symbol: Ldhal6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175349.2 | NP_780558.1 | 1 | 1030 | ||
GenBank mRNA | AK077089.1 | - | 1 | 976 | ||
AY280363.1 | - | 1 | 1030 | |||
BC061193.1 | - | 1 | 975 | 6704800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
