siRNA Details
Gene Symbol: Itfg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206335.1 | NP_001193264.1 | 3 | 631 | ||
NM_207217.4 | NP_997100.1 | 2 | 519 | |||
GenBank mRNA | AK029427.1 | - | 2 | 419 | ||
AK158049.1 | - | 613 | ||||
AK170646.1 | - | 3 | 550 | |||
BC046457.1 | - | 3 | 478 | 4222961 |
