siRNA Details
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134122.2 | NP_598883.1 | 3 | 525 | ||
GenBank mRNA | BC021750.1 | - | 3 | 528 | 4503599 | |
BC026576.1 | - | 3 | 497 | 4984796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
