siRNA Details
Gene Symbol: Tcf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163763.1 | NP_001157235.1 | 3 | 917 | ||
NM_001163764.1 | NP_001157236.1 | 3 | 760 | |||
NM_025674.2 | NP_079950.2 | 3 | 808 | |||
GenBank mRNA | AK077414.1 | - | 3 | 796 | ||
AK078697.1 | - | 3 | 880 | |||
AK088479.1 | - | 3 | 772 | |||
BC004617.1 | - | 3 | 738 | 3585301 | ||
BG082313.2 | - | 2 | 398 | |||
S53376.1 | - | 3 | 733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
