siRNA Details
Gene Symbol: Kctd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142731.1 | NP_001136203.1 | 3, 4 | 2144 | ||
NM_134112.5 | NP_598873.2 | 3, 4 | 460 | |||
GenBank mRNA | AK016623.1 | - | 2, 3 | 505 | ||
AY899808.1 | - | 3, 4 | 460 | |||
BC022615.1 | - | 1, 2 | 128 | |||
BC087623.1 | - | 3, 4 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
