siRNA Details
Gene Symbol: Slc39a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134135.1 | NP_598896.1 | 2 | 1881 | ||
GenBank mRNA | AK087519.1 | - | 4 | 2039 | ||
AK205681.1 | - | 1 | 171 | |||
BC005502.1 | - | 2 | 1881 | 3490154 | ||
BC085175.1 | - | 4 | 2019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
