siRNA Details
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134136.3 | NP_598897.2 | 18 | 3096 | ||
GenBank mRNA | AK031096.1 | - | 18 | 3046 | ||
AK031347.1 | - | 18 | 3027 | |||
AK089046.1 | - | 18 | 3065 | |||
AY267463.1 | - | 18 | 3026 | |||
BC056348.1 | - | 18 | 3048 | 5721407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
