siRNA Details
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134136.3 | NP_598897.2 | 15 | 2112 | ||
GenBank mRNA | AK031096.1 | - | 15 | 2062 | ||
AK031347.1 | - | 15 | 2043 | |||
AK034761.1 | - | 10 | 1299 | |||
AK089046.1 | - | 15 | 2084 | |||
AY267463.1 | - | 15 | 2045 | |||
BC056348.1 | - | 15 | 2064 | 5721407 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006254825, XM_006254820, XM_006254821, XM_006254819, XM_006254822, and others.
Gene Ontology Categories:
