siRNA Details
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134139.1 | NP_598900.1 | 7, 8 | 725 | ||
GenBank mRNA | AK077521.1 | - | 7, 8 | 726 | ||
AK145704.1 | - | 7, 8 | 606 | |||
BC019968.1 | - | 7, 8 | 725 | 4167260 | ||
BC024478.1 | - | 3, 4 | 320 | 4977054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
