siRNA Details
Gene Symbol: Uqcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160356.1 | NP_001153828.1 | 2 | 596 | ||
GenBank mRNA | AK131602.1 | - | 2 | 587 | ||
AK131892.1 | - | 2 | 560 | |||
AK132116.1 | - | 2 | 587 | |||
AY941793.1 | - | 2 | 586 |
