siRNA Details
Gene Symbol: Ffar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181748.2 | NP_861413.1 | 2 | 718 | ||
GenBank mRNA | AB115769.1 | - | 2 | 670 | ||
AK131616.1 | - | 2 | 705 | |||
AY288424.1 | - | 2 | 670 | |||
BC053698.1 | - | 2 | 718 | 6529120 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
