siRNA Details
Gene Symbol: Macrod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134147.4 | NP_598908.2 | 3 | 570 | ||
GenBank mRNA | AK052998.1 | - | 3 | 529 | ||
BC008653.1 | - | 3 | 295 | 3597023 | ||
BQ126199.1 | - | 3 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
