siRNA Details
Gene Symbol: Kazald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178929.4 | NP_849260.2 | 5 | 1002 | ||
GenBank mRNA | AK030678.1 | - | 5 | 1001 | ||
AK155095.1 | - | 6 | 1247 | |||
BC115642.1 | - | 5 | 993 | 40103018 | ||
BC115643.1 | - | 5 | 985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
