siRNA Details
Gene Symbol: Exoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175353.2 | NP_780562.2 | 16 | 1647 | ||
GenBank mRNA | AK082881.1 | - | 16 | 1626 | ||
AK151524.1 | - | 16 | 1648 | |||
AK151838.1 | - | 16 | 1648 | |||
BC025616.1 | - | 3 | 221 | |||
BC026859.1 | - | 16 | 1600 | 2631201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
