siRNA Details
Gene Symbol: Slc25a45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134154.3 | NP_598915.1 | 2, 3 | 279 | ||
GenBank mRNA | AK133879.1 | - | 2, 3 | 933 | ||
AK155754.1 | - | 2, 3 | 932 | |||
AK169876.1 | - | 2, 3 | 925 | |||
AK170869.1 | - | 3, 4 | 410 | |||
BC022156.1 | - | 2, 3 | 110 | |||
BC037680.1 | - | 2, 3 | 283 | 3481846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
