siRNA Details
Gene Symbol: Unc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029770.2 | NP_084046.2 | 9 | 1602 | ||
GenBank mRNA | AJ487853.1 | - | 9 | 1505 | ||
AK147603.1 | - | 8 | 1464 | |||
AK154271.1 | - | 9 | 1602 | |||
AK154287.1 | - | 9 | 1605 | |||
AK158284.1 | - | 9 | 1602 | |||
AK170513.1 | - | 9 | 1601 | |||
BC048162.1 | - | 8 | 1376 | |||
BC057560.1 | - | 8 | 1376 | 6417563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
