siRNA Details
Gene Symbol: Mkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181860.1 | NP_862908.1 | 3, 4 | 333 | ||
GenBank mRNA | BC028459.1 | - | 3, 4 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
