siRNA Details
Gene Symbol: Guca1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146079.1 | NP_666191.1 | 2, 3 | 460 | ||
GenBank mRNA | AK033052.1 | - | 2, 3 | 425 | ||
BC018258.1 | - | 2, 3 | 388 | 4511650 | ||
BC018480.1 | - | 2, 3 | 429 | 4500919 | ||
BC029017.1 | - | 2, 3 | 405 | 5359531 | ||
BC029661.1 | - | 2, 3 | 415 | 5366068 | ||
BC031367.1 | - | 2, 3 | 423 | 5358667 | ||
BC031916.1 | - | 2, 3 | 460 | 5369642 | ||
BC033348.1 | - | 2, 3 | 410 | |||
BC033358.1 | - | 2, 3 | 459 | 5368968 | ||
BC033364.1 | - | 2, 3 | 410 | |||
BC033380.1 | - | 2, 3 | 415 | 5366953 | ||
BC033383.1 | - | 2, 3 | 416 | 5368064 | ||
BC033396.1 | - | 2, 3 | 407 | |||
BC033423.1 | - | 2, 3 | 412 | |||
BC034331.1 | - | 2, 3 | 416 | 5357797 | ||
BC034595.1 | - | 2, 3 | 415 | 5362191 |
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027134.1 | - | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
