siRNA Details
Gene Symbol: Atf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030693.2 | NP_109618.1 | 3 | 1095 | ||
GenBank mRNA | AB012276.1 | - | 1 | 205 | ||
AF310624.1 | - | 3 | 1116 | |||
AF312938.1 | - | 3 | 1071 | |||
AF375475.1 | - | 3 | 1100 | |||
AF375476.1 | - | 3 | 1095 | |||
AK131939.1 | - | 3 | 1117 | |||
AK146479.1 | - | 3 | 1117 | |||
AK159624.1 | - | 3 | 1119 | |||
AK160085.1 | - | 1 | 520 | |||
AY024362.1 | - | 1 | 610 | |||
BC010195.1 | - | 2 | 928 | 3157499 | ||
BC092068.1 | - | 3 | 1100 | 4210931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
