siRNA Details
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139293.2 | NP_647454.2 | 5, 6 | 763 | ||
NM_177940.1 | NP_808809.1 | 5, 6 | 983 | |||
NM_177941.1 | NP_808810.1 | 6, 7 | 1070 | |||
NM_177942.1 | NP_808811.1 | 4, 5 | 676 | |||
GenBank mRNA | AF396699.1 | - | 5, 6 | 704 | ||
AF489569.1 | - | 5, 6 | 802 | |||
AF489570.1 | - | 6, 7 | 889 | |||
AF489571.1 | - | 4, 5 | 676 | |||
AF489572.1 | - | 5, 6 | 763 | |||
AK031951.1 | - | 5, 6 | 771 | |||
AK043473.1 | - | 3, 4 | 426 | |||
BC138052.1 | - | 5, 6 | 670 | 8861071 | ||
BC138053.1 | - | 5, 6 | 670 | 8861072 | ||
BC145126.1 | - | 5, 6 | 670 |
