siRNA Details
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139293.2 | NP_647454.2 | 1, 2 | 201 | ||
GenBank mRNA | AF396699.1 | - | 1, 2 | 142 | ||
AF489572.1 | - | 1, 2 | 201 | |||
AK031951.1 | - | 1, 2 | 209 | |||
AU080537.1 | - | 1, 2 | 158 | |||
BC138052.1 | - | 1, 2 | 108 | 8861071 | ||
BC138053.1 | - | 1, 2 | 108 | 8861072 | ||
BC145126.1 | - | 1, 2 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
