siRNA Details
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139293.2 | NP_647454.2 | 7 | 941 | ||
NM_177940.1 | NP_808809.1 | 7 | 1161 | |||
NM_177941.1 | NP_808810.1 | 8 | 1248 | |||
NM_177942.1 | NP_808811.1 | 6 | 854 | |||
GenBank mRNA | AF396699.1 | - | 7 | 882 | ||
AF489569.1 | - | 7 | 980 | |||
AF489570.1 | - | 8 | 1067 | |||
AF489571.1 | - | 6 | 854 | |||
AF489572.1 | - | 7 | 941 | |||
AK031951.1 | - | 7 | 949 | |||
AK043473.1 | - | 5 | 604 | |||
BC030900.1 | - | 2 | 64 | |||
BC138052.1 | - | 7 | 848 | 8861071 | ||
BC138053.1 | - | 7 | 848 | 8861072 | ||
BC145126.1 | - | 7 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
