siRNA Details
Gene Symbol: Dio3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172119.2 | NP_742117.2 | 1 | 306 | ||
GenBank mRNA | AK140797.1 | - | 1 | 307 | ||
AK146060.1 | - | 1 | 307 | |||
BC106847.1 | - | 1 | 276 | 40046057 | ||
BC106848.1 | - | 1 | 276 | 40046058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
