siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 31 | 5594 | ||
GenBank mRNA | AF314149.1 | - | 2 | 419 | ||
AK078556.1 | - | 12 | 2271 | |||
AK162008.1 | - | 13 | 1957 | |||
AY237726.1 | - | 31 | 5623 | |||
AY237727.2 | - | 16 | 3147 | |||
BC034209.1 | - | 6 | 741 | |||
BC037721.1 | - | 16 | 2157 | |||
BC038902.1 | - | 2 | 367 | |||
BC045197.1 | - | 16 | 3077 | |||
BC145279.1 | - | 31 | 5581 | 9053833 | ||
BC150749.1 | - | 31 | 5581 | 9087660 | ||
BC171954.1 | - | 31 | 5582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
