siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 31 | 5580 | ||
GenBank mRNA | AF314149.1 | - | 2 | 405 | ||
AK078556.1 | - | 12 | 2257 | |||
AK162008.1 | - | 13 | 1943 | |||
AY237726.1 | - | 31 | 5609 | |||
AY237727.2 | - | 16 | 3133 | |||
BC034209.1 | - | 6 | 727 | |||
BC037721.1 | - | 16 | 2143 | |||
BC038902.1 | - | 2 | 353 | |||
BC045197.1 | - | 16 | 3063 | |||
BC058610.1 | - | 31 | 5592 | 30056142 | ||
BC145279.1 | - | 31 | 5567 | 9053833 | ||
BC150749.1 | - | 31 | 5567 | 9087660 | ||
BC171954.1 | - | 31 | 5568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
