siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 22 | 4312 | ||
GenBank mRNA | AK162008.1 | - | 4 | 675 | ||
BC030478.1 | - | 12 | 2544 | |||
BC037721.1 | - | 7 | 875 | |||
BC045197.1 | - | 7 | 1795 | |||
BC058610.1 | - | 22 | 4324 | 30056142 | ||
BC145279.1 | - | 22 | 4299 | 9053833 | ||
BC150749.1 | - | 22 | 4299 | 9087660 | ||
BC171954.1 | - | 22 | 4300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
