siRNA Details
Gene Symbol: Nod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171007.1 | NP_001164478.1 | 3, 4 | 582 | ||
NM_172729.3 | NP_766317.1 | 3, 4 | 833 | |||
GenBank mRNA | AK089662.1 | - | 3, 4 | 583 | ||
AK137585.1 | - | 3, 4 | 838 | |||
AY160222.1 | - | 2, 3 | 368 | |||
BC042670.1 | - | 3, 4 | 660 | 3597911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
