siRNA Details
Gene Symbol: Uap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036243.1 | - | 2 | 367 | ||
AK047566.1 | - | 2 | 432 | |||
AK049519.1 | - | 2 | 421 | |||
AK081690.1 | - | 2 | 392 | |||
AK147139.1 | - | 2 | 367 | |||
AK163314.1 | - | 2 | 266 | |||
AK171925.1 | - | 2 | 264 | |||
AK218390.1 | - | 1 | 152 | |||
BC016406.1 | - | 2 | 354 | 4489112 | ||
BC017547.1 | - | 2 | 243 | 4920702 | ||
BM947026.1 | - | 2 | 439 |
