siRNA Details
Gene Symbol: Mrpl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031808.2 | NP_001026978.2 | 2 | 552 | ||
GenBank mRNA | AK003340.1 | - | 2 | 467 | ||
AK011837.1 | - | 2 | 491 | |||
AK032370.1 | - | 2 | 552 | |||
BC034728.1 | - | 2 | 491 | 2811521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
