siRNA Details
Gene Symbol: Prrxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001796.4 | NP_001001796.1 | 6 | 699 | ||
GenBank mRNA | AK039633.1 | - | 6 | 700 | ||
BC132613.1 | - | 6 | 668 | 40130890 | ||
BC145917.1 | - | 6 | 668 | 40131220 | ||
EU670678.1 | - | 7 | 819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
