siRNA Details
Gene Symbol: Haao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025325.2 | NP_079601.1 | 2 | 208 | ||
GenBank mRNA | AK002295.1 | - | 2 | 230 | ||
AK002608.1 | - | 2 | 168 | |||
AK171228.1 | - | 3 | 1362 | |||
BC012872.1 | - | 2 | 208 | 4196161 | ||
BC094021.1 | - | 2 | 208 | 4163839 |

More Information
Other Species Targeted

Gene Ontology Categories:
