siRNA Details
Gene Symbol: Bmyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023326.2 | NP_075815.2 | 1 | 731 | ||
GenBank mRNA | AK012810.1 | - | 1 | 773 | ||
AK013473.1 | - | 1 | 781 | |||
AK013780.1 | - | 1 | 777 | |||
AK158603.1 | - | 1 | 781 | |||
BC056436.1 | - | 1 | 731 | 6824373 | ||
BC060207.1 | - | 1 | 731 | 6824373 | ||
BC061001.1 | - | 1 | 830 | 6595436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
