siRNA Details
Gene Symbol: Mthfs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026829.2 | NP_081105.1 | 2 | 201 | ||
GenBank mRNA | AK004092.1 | - | 2 | 202 | ||
AK043235.1 | - | 2 | 178 | |||
AK154942.1 | - | 3 | 874 | |||
AK158264.1 | - | 210 |
Gene Symbol: Mthfsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128601.1 | NP_001122073.1 | 2 | 219 | ||
GenBank mRNA | AK149809.1 | - | 1 | 390 | ||
AK149842.1 | - | 1 | 390 | |||
BC024567.1 | - | 2 | 111 | 5026828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
