siRNA Details
Gene Symbol: Frs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144939.2 | NP_659188.1 | 5 | 550 | ||
GenBank mRNA | BC014819.1 | - | 5 | 550 | 4506982 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
