siRNA Details
Gene Symbol: Gtpbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145147.5 | NP_660129.2 | 7 | 1038 | ||
GenBank mRNA | AJ293619.1 | - | 6 | 668 | ||
AK082192.1 | - | 4 | 914 | |||
AK144812.1 | - | 7 | 1039 | |||
AK152225.1 | - | 7 | 1039 | |||
AK152972.1 | - | 7 | 1039 | |||
AK159462.1 | - | 7 | 1011 | |||
BC027821.1 | - | 4 | 569 | 5321725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
