siRNA Details
Gene Symbol: Cenpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081363.2 | NP_001074832.2 | 12 | 3885 | ||
GenBank mRNA | AF194970.1 | - | 1 | 2170 | ||
DQ642022.1 | - | 11 | 3742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
