siRNA Details
Gene Symbol: Ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290378.1 | NP_001277307.1 | 3 | 526 | ||
NM_001290379.1 | NP_001277308.1 | 3 | 526 | |||
NM_026887.3 | NP_081163.2 | 3 | 526 | |||
GenBank mRNA | AK005223.1 | - | 3 | 390 | ||
AK045558.1 | - | 3 | 527 | |||
AK047724.1 | - | 3 | 403 | |||
AK053941.1 | - | 3 | 399 | |||
AK167690.1 | - | 3 | 401 | |||
AK211598.1 | - | 1 | 12 | |||
BC046964.1 | - | 3 | 383 | 6485320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
