siRNA Details
Gene Symbol: Fxyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173372.1 | NP_001166843.1 | 7 | 306 | ||
NM_033648.1 | NP_387468.1 | 7 | 309 | |||
GenBank mRNA | AK018728.1 | - | 7 | 306 | ||
BC086918.1 | - | 8 | 570 | 6762406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
