siRNA Details
Gene Symbol: Rnf144a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081977.2 | NP_001075446.1 | 6 | 682 | ||
NM_080563.4 | NP_542130.1 | 5 | 608 | |||
GenBank mRNA | AF360998.1 | - | 6 | 678 | ||
AK038586.1 | - | 5 | 500 | |||
AK133544.1 | - | 3 | 222 | |||
AK172904.1 | - | 4 | 443 | |||
BC030187.1 | - | 5 | 572 | 5345434 | ||
BQ884505.1 | - | 5 | 541 | |||
BU055293.1 | - | 7 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
