siRNA Details
Gene Symbol: Med21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025315.3 | NP_079591.1 | 2 | 136 | ||
GenBank mRNA | AK009742.1 | - | 2 | 137 | ||
AK012217.1 | - | 2 | 137 | |||
AK014428.1 | - | 2 | 130 | |||
AK019417.1 | - | 2 | 126 | |||
BC012286.1 | - | 2 | 127 | 3493056 | ||
BC096641.1 | - | 1 | 1086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
