siRNA Details
Gene Symbol: Slc22a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144856.2 | NP_659105.2 | 6 | 1084 | ||
GenBank mRNA | AB069965.1 | - | 6 | 996 | ||
AK082865.1 | - | 6 | 1085 | |||
AK143949.1 | - | 6 | 1072 | |||
BC013474.1 | - | 6 | 1061 | 4236556 | ||
BC024119.1 | - | 6 | 1048 | 5096875 | ||
BC025813.1 | - | 6 | 715 | 5097639 | ||
BC026597.1 | - | 6 | 1038 | 4975360 | ||
BC026598.1 | - | 6 | 1036 | 4975598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
