siRNA Details
Gene Symbol: Xrcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028012.4 | NP_082288.1 | 2 | 219 | ||
GenBank mRNA | AB055154.1 | - | 2 | 127 | ||
AK009951.1 | - | 2 | 136 | |||
AK053612.1 | - | 135 | ||||
AK088281.1 | - | 2 | 135 | |||
BC025538.1 | - | 2 | 163 | 5257949 | ||
BY774472.1 | - | 2 | 219 |
