siRNA Details
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197321.1 | NP_001184250.1 | 15 | 1852 | ||
NM_001197322.1 | NP_001184251.1 | 12 | 1459 | |||
NM_053202.2 | NP_444432.1 | 16 | 2004 | |||
GenBank mRNA | AB221601.1 | - | 13 | 1695 | ||
AB221602.1 | - | 13 | 1697 | |||
AB594192.1 | - | 17 | 2409 | |||
AF339103.1 | - | 13 | 1693 | |||
AF339105.1 | - | 1473 | ||||
AK033368.1 | - | 10 | 1169 | |||
AK077062.1 | - | 12 | 1354 | |||
AK160315.1 | - | 4 | 456 | |||
BC064764.1 | - | 15 | 1838 | 30430449 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001034131, XM_008763154, XM_008763152, XM_008763148, XM_008763150, and others.
Gene Ontology Categories:
