siRNA Details
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197321.1 | NP_001184250.1 | 14 | 1704 | ||
NM_001197322.1 | NP_001184251.1 | 11 | 1311 | |||
NM_053202.2 | NP_444432.1 | 15 | 1856 | |||
GenBank mRNA | AB221601.1 | - | 12 | 1548 | ||
AB221602.1 | - | 12 | 1549 | |||
AB594192.1 | - | 16 | 2261 | |||
AF339103.1 | - | 12 | 1545 | |||
AF339104.1 | - | 12 | 1545 | |||
AF339105.1 | - | 1325 | ||||
AK033368.1 | - | 9 | 1021 | |||
AK077062.1 | - | 11 | 1206 | |||
AK160315.1 | - | 3 | 308 | |||
BC064764.1 | - | 14 | 1690 | 30430449 |

More Information
Other Species Targeted

Gene Ontology Categories:
