siRNA Details
Gene Symbol: Fam207a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133998.3 | NP_598759.1 | 3 | 379 | ||
GenBank mRNA | AF391115.1 | - | 3 | 352 | ||
AK076516.1 | - | 3 | 350 | |||
AK088306.1 | - | 3 | 347 | |||
AK160157.1 | - | 3 | 250 | |||
BB840302.1 | - | 3 | 380 | |||
BC034216.1 | - | 3 | 325 | 5008732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
