siRNA Details
Gene Symbol: Sft2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145512.4 | NP_663487.1 | 6, 7 | 548 | ||
GenBank mRNA | AK036578.1 | - | 6, 7 | 436 | ||
AK075812.1 | - | 5, 6 | 378 | |||
AK156973.1 | - | 6, 7 | 439 | |||
AK170590.1 | - | 5, 6 | 353 | |||
BC017549.1 | - | 6, 7 | 427 | 4924681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
