siRNA Details
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133985.2 | NP_598746.2 | 17 | 1841 | ||
GenBank mRNA | AK031790.1 | - | 16 | 1338 | ||
AK075837.1 | - | 16 | 1349 | |||
AK129291.1 | - | 17 | 1836 | |||
BC036148.1 | - | 8 | 591 | |||
BC046453.1 | - | 18 | 1865 | |||
BC060645.1 | - | 17 | 1824 | 6852021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
