siRNA Details
Gene Symbol: Lyrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175364.4 | NP_780573.1 | 3 | 234 | ||
GenBank mRNA | AK044791.1 | - | 3 | 229 | ||
BC028533.1 | - | 3 | 216 | 1446940 | ||
BY131987.1 | - | 3 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
