siRNA Details
Gene Symbol: Pnrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033225.2 | NP_001028397.2 | 2 | 1094 | ||
GenBank mRNA | AK087995.1 | - | 2 | 819 | ||
AK159693.1 | - | 2 | 817 | |||
AK160284.1 | - | 2 | 818 | |||
BC038496.1 | - | 1 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
