siRNA Details
Gene Symbol: Rcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173867.5 | NP_776292.1 | 3 | 546 | ||
GenBank mRNA | AK076040.1 | - | 3 | 477 | ||
AK076046.1 | - | 777 | ||||
AK129370.1 | - | 2 | 250 | |||
BC020385.1 | - | 1 | 79 | |||
BC060624.1 | - | 3 | 500 | |||
BC086666.1 | - | 3 | 469 | 30634817 | ||
CA751510.1 | - | 3 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
